Learn More
Description
- Tcirg1 T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3 [ Mus musculus (house mouse) ]
- Also known as: oc; Stv1; Vph1; Atp6i; OPTB1; TIRC7; ATP6a3; OC116; ATP6N1C
- The TCIRG1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- TCIRG1 is a subunit of a large protein complex known as a vacuolar H+-ATPase (V-ATPase).
- The protein complex acts as a pump to move protons across the membrane.
- This movement of protons helps regulate the pH of cells and their surrounding environment.
- V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptormediated endocytosis.
- Mutations in TCORG1 are associated with infantile malignant osteopetrosis.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mTcirg1 |
| Accession Number | NP_058617 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2518 bp |
| Gene ID (Entrez) | 10312 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
