Learn More
Sino Biological Mouse TEK Protein 20UG 51087-M08H-B-20
Supplier:  Sino Biological 51087M08HB20
Encompass_Preferred
A DNA sequence encoding the Mouse TEK (Q02858) (Met1-Lys744) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-240-9832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.