Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse TFF-1 Recombinant Protein, PeproTech®

Catalog No. 31531100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31531100UG 100 μg
315-31-1MG 1 mg
315-31-20UG 20 μg
31531250UG 250 μg
31531500UG 500 μg
315-31-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 31531100UG Supplier Gibco™ Supplier No. 31531100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine TFF-1 is a 7.4 kDa monomeric protein consisting of a 66 amino acid polypeptide chain, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08423
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 21784
Molecular Weight (g/mol) 7.4 kDa
Name Mouse TFF-1
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1
Common Name TFF1
Gene Symbol TFF1
Conjugate Unconjugated
Recombinant Recombinant
Sequence QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.