Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse TFF-2 Recombinant Protein, PeproTech®

Catalog No. 3153020UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-30-20UG 20 μg
31530100UG 100 μg
315-30-1MG 1 mg
31530250UG 250 μg
31530500UG 500 μg
315-30-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 315-30-20UG Supplier Gibco™ Supplier No. 3153020UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine TFF-2 is an 11.9 kDa polypeptide of 106 amino acid residues, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing and protection of the gastrointestinal epithelia. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain. SP (spasmolytic polypeptide), also designated Trefoil factor 2 (TFF2) precursor, is a trefoil protein that functions to inhibit gastrointestinal motility and gastric acid secretion. SP may also act as a structural component of the gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains. A downregulation of SP expression is associated with primary gastric cancer, and a progressive loss of this protein is likely to be involved in the early stage of the multi-step gastric carcinogenesis pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03404
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 21785
Molecular Weight (g/mol) 11.9 kDa
Name Mouse TFF-2
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1)
Common Name TFF2
Gene Symbol Tff2
Conjugate Unconjugated
Recombinant Recombinant
Sequence EKPSPCRCSR LTPHNRKNCG FPGITSEQCF DLGCCFDSSV AGVPWCFHPL PNQESEQCVM EVSARKNCGY PGISPEDCAS RNCCFSNLIF EVPWCFFPQS VEDCHY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.