Learn More
Sino Biological Protein Mouse TFPI 50ug
Supplier:  Sino Biological 50131M08H50
Encompass_Preferred
A DNA sequence encoding the mouse TFPI isoform alpha (NP 035706 1) (Met 1-Lys 289) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-224-7265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.