Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse TGF-alpha ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349958 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349958 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349958 Supplier Novus Biologicals™ Supplier No. NBP349958
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 7.81 to 500 pg/mL
Assay Sensitivity 4.69 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Mouse TGF-alpha in samples. No significant cross-reactivity or interference between Mouse TGF-alpha and analogues was observed
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 7039
Gene Symbol TGFA
Quantity 1 Kit
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.