Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse TGF-beta 1 DuoSet ELISA

Catalog No. p-4754877
Change view
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DY1679 15 Plates
DY167905 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY1679 Supplier R&D Systems™ Supplier No. DY1679
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse TGF-beta 1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse TGF-beta 1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TGF-beta 1.

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit
Species Mouse
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 21803
Synonym CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.