Learn More
Bon Opus Biosciences Recombinant Mouse TGFBR2 (N-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107450UG
Encompass_Preferred
Recombinant Mouse TGFBR2 (N-hFc) is produced by Human Cells. The target gene encoding Ile24-Asp184 is expressed with a N-hFc tag. Accession# Q62312. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1074
Catalog No. 50-217-2672
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.