Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Thrombopoietin Quantikine ELISA Kit
SDP

Catalog No. MTP00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 x 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTP00 2 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MTP00 Supplier R&D Systems™ Supplier No. MTP00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Thrombopoietin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Thrombopoietin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Thrombopoietin/THPO in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), citrate plasma (10 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 20pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum
Specificity Natural and Recombinant Mouse Tpo
For Use With (Application) ELISA
Gene ID (Entrez) 21832
Quantity 2 x 96-well Plates
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Citrate Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.