Learn More
American Research Products Inc Mouse Thyroid Stimulating Horm
Supplier:  American Research Products Inc CSBE05116M
Encompass
Mouse Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 3 IU/ml-12 IU/ml Sensitivity 0 15 IU/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.