Learn More
ACROBiosystems Mouse TIGIT 25ug
Supplier:  ACROBiosystems TITM82E925UG
Encompass_Preferred
Biotinylated Mouse TIGIT Protein HisAvitag (TIT-M82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # A0A0B4J1G6-1).
Catalog No. 50-253-4049
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.