Learn More
Description
Specifications
Specifications
| Assay Range | 7.81 - 500pg/mL |
| Assay Sensitivity | 2.18pg/mL |
| Conjugate | HRP |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Mouse TIM-1. The antibody components of this kit were raised against the short splice form of Mouse TIM-1 (isoform 2) which has a 23 amino acid deletion following Pro182. This immunoassay detects both the short and long forms of Mouse TIM-1. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 171283 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
