Learn More
Kingfisher Biotech Inc Mouse TIM-3 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1470M005
Encompass
The Mouse TIM-3 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse TIM-3 applications are for cell culture, ELISA standard, and Western Blot Control. Mouse TIM-3 yeast-derived recombinant protein can be purchased in multiple sizes. Mouse TIM-3 Specifications: (Molecular Weight: 19.3 kDa) (Amino Acid Sequence: RSLENAYVFE VGKNAYLPCS YTLSTPGALV PMCWGKGFCP WSQCTNELLR TDERNVTYQK SSRYQLKGDL NKGDVSLIIK NVTLDDHGTY CCRIQFPGLM NDKKLELKLD IKAAKVTPAQ TAHGDSTTAS PRTLTTERNG SETQTLVTLH NNNGTKISTW ADEIKDSGET IRTA (174)) (Gene ID: 171285). For research use only. Made in the USA
Catalog No. 50-253-3241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.