Learn More
R&D Systems™ Mouse TNF-alpha Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 10.90 - 700pg/mL |
| Assay Sensitivity | 7.21pg/mL |
| Conjugate | HRP |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Mouse TNF-alpha |
| Sufficient For | 2 x 96-well Plates |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 21926 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.