Learn More
Description
- Also known as: Tnfrh3; Tnfrsf24
- TNFRSF26 is a member of the TNF receptor superfamily.
- It appears to be concentrated in lymphoid cell types.
- Unlike two related TNF receptor homologs, TNFRSF26 does not serve as a TRAIL decoy receptor.
- No ortholog to mouse TNFRSF26 has been reported in human or rat.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mTnfrsf26 |
| Accession Number | NP_783580 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 628 bp |
| Gene ID (Entrez) | 8718 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
