Learn More
Sino Biological Mouse TNFSF9 MAb 100TESTS 50067-R014-C-100
Supplier:  Sino Biological 50067R014C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse 4-1BBL / CD137L / TNFSF9 (rM 4-1BBL / CD137L / TNFSF9; Catalog#50067-M07H; NP_033430.1; Arg 104-Glu 309) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-2505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.