Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Total MMP-9 Quantikine ELISA Kit
SDP

Catalog No. MMPT90 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMPT90 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MMPT90 Supplier R&D Systems™ Supplier No. MMPT90
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Total MMP-9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Total MMP-9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse MMP-9 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.08 to 5 ng/mL
Assay Sensitivity 0.014 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Plasma (Heparin), Serum, Serum
Specificity Natural and Recombinant Mouse MMP-9 (pro-, active, and TIMP-complexed)
For Use With (Application) ELISA
Gene ID (Entrez) 4318
Gene Symbol Mmp9
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Target MMP-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.