Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse TPO (Thrombopoietin) Recombinant Protein, PeproTech®

Catalog No. 315142UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-14-2UG 2 μg
31514100UG 100 μg
315-14-10UG 10 μg
315-14-1MG 1 mg
31514250UG 250 μg
31514500UG 500 μg
315-14-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 315-14-2UG Supplier Gibco™ Supplier No. 315142UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40226
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 21832
Molecular Weight (g/mol) 18.7 kDa
Name Mouse TPO (Thrombopoietin)
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-mpl ligand; Megakaryocyte colony-stimulating factor; megakaryocyte growth and development factor; megakaryocyte stimulating factor; Mgdf; MKCSF; ML; MPL ligand; Mpllg; MSA; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; TDH2A; THCYT1; THPO; Thrombopoietin; thyroid microsomal antigen; thyroid peroxidase; thyroperoxidase; Tpo; TPX
Common Name Thrombopoietin
Gene Symbol THPO
Biological Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of human MO7e cells was found to be ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence SPVAPACDPR LLNKLLRDSH LLHSRLSQCP DVDPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPLQGRTTA HKDPNALFLS LQQLLRGKVR FLLLVEGPTL CVRRTLPTTA VPSSTSQLLT LNKF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.