Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse TRACP/PAP/ACP5 ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Description
The Mouse TRACP/PAP/ACP5 ELISA Kit (Chemiluminescence) measures mouse TRACP/PAP/ACP5 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 0.156 to 10 ng/mL |
| Assay Sensitivity | 0.094 ng/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Mouse TRACP-5b in samples. No significant cross-reactivity or interference between Mouse TRACP-5b and analogues was observed. |
| Gene ID (Entrez) | 54 |
| Gene Symbol | ACP5 |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?