Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TREM-1 Alexa Fluor Plus 594-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10138746
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10138746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10138746 Supplier R&D Systems Supplier No. AF1187AFP594100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

TREM1 Polyclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry,Western Blot,Agonist Activity,ELISA Capture (Matched Antibody Pair),Immunocytochemistry,CyTOF

Specifications

Antigen TREM1
Applications Flow Cytometry, Western Blot, Agonist Activity, ELISA Capture (Matched Antibody Pair), Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor Plus 594
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54210
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Product Type Antibody
Form Liquid
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.