Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse TREM2 (NP_001259007) VersaClone cDNA
SDP

Catalog No. RDC2449 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2449 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2449 Supplier R&D Systems™ Supplier No. RDC2449

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Mouse TREM-2 (NP_001259007) VersaClone cDNA clone contains the complete ORF for TREM-2, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The TREM-2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided

Specifications

Gene mTREM-2
Accession Number NP_001259007
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 54209
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2 triggering receptor expressed on myeloid cells 2 A, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Content And Storage Store the unopened product at -20 to -80°C
Species Mouse
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.