Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Tryptase epsilon /BSSP-4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10006728
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10006728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10006728 Supplier R&D Systems Supplier No. FAB20592R100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Tryptase epsilon/BSSP-4/PRSS22 Monoclonal antibody specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)

Specifications

Antigen Tryptase epsilon/BSSP-4/PRSS22
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 246702
Conjugate Alexa Fluor 647
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tryptase Îμ/BSSP‑4, Ala33-Ser306, Accession # Q9ER10
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64063
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.