Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TWEAK/TNFSF12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1237 Supplier R&D Systems Supplier No. AF1237
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen TWEAK/TNFSF12
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 4-16 ug/mL
Gene Accession No. O54907
Gene Alias APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
Gene Symbols Tnfsf12
Host Species Goat
Immunogen E. coli-derived recombinant mouse TWEAK/TNFSF12 Arg105-His249 Accession # O54907
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8742
Test Specificity Detects TWEAK/TNFSF12 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human TWEAK is observed and less than 1% cross-reactivity with recombinant mouse (rm) BAFF, rmFas Ligand, rmOX40 Ligand, rmTNF-alpha , and rmTRAIL is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.