Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Tyro3/Dtk Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB759SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB759SP 25 μg
MAB759 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB759SP Supplier R&D Systems Supplier No. MAB759SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 6 publications

Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Dtk/TYRO3
Applications Western Blot, Immunohistochemistry, CyTOF, ELISA
Classification Monoclonal
Clone 109646
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P55144
Gene Alias Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Gene Symbols Tyro3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7301
Test Specificity Detects mouse Tyro3/Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Tyro3/Dtk, recombinant mouse (rm) Mer, or rmAxl is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.