Learn More
Description
- Also known as: CD106; Vcam-1
- VCAM1 is a member of the Ig superfamily and is a cell surface sialoglycoprotein expressed by cytokine-activated endothelium.
- It is a type I membrane protein that mediates leukocyte-endothelial cell adhesion and signal transduction and may play a role in the development of artherosclerosis and rheumatoid arthritis.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mVcam1 |
| Accession Number | AAA40545 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2233 bp |
| Gene ID (Entrez) | 7412 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
