Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VE-Cadherin Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. AF1002
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1002 Supplier R&D Systems Supplier No. AF1002
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

VE-Cadherin Polyclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen VE-Cadherin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL
Gene Accession No. 2208309A
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin
Gene Symbols CDH5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 Accession # 2208309A
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1003
Test Specificity Detects mouse VE-Cadherin in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human VE-Cadherin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.