Learn More
Mouse VE-Statin Isoform 1 Antibody, R&D Systems™



Description
Specifications
Specifications
| Antigen | VE-Statin/EGFL7 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q9QXT5 |
| Gene Alias | EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 |
| Gene Symbols | EGFL7 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.