Learn More
Gibco™ Mouse VEGF-165 Recombinant Protein, PeproTech®
Description
Recombinant Murine VEGF is a 39.0 kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q00731 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 22339 |
| Molecular Weight (g/mol) | 39 kDa |
| Name | Mouse VEGF-165 |
| Quantity | 10 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.