Learn More
Proteintech Group Inc Mouse VEGF ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE1000996T
Encompass_Preferred
Vascular endothelial growth factor (VEGF), is a signal protein produced by cells that stimulates vasculogenesis and angiogenesis. It is part of the system that restores the oxygen supply to tissues when blood circulation is inadequate such as in hypoxic conditions. Serum concentration of VEGF is high in bronchial asthma and diabetes mellitus. The activities of VEGF are not limited to the vascular system; VEGF plays a role in normal physiological functions such as bone formation, hematopoiesis, wound healing, and development. Disruption of this gene in mice resulted in abnormal embryonic blood vessel formation. VEGF is upregulated in many known tumors and its expression is correlated with tumor stage and progression.
Catalog No. 50-197-4135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.