Learn More
Description
Specifications
Specifications
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 20.0pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Specificity | This assay detects both natural and recombinant mouse VEGF-A. The cross reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a mouse VEGF-A positive serum. There was no cross reactivity or interference detected, notably not with mouse VEGF-B, VEGF-C, VEGF-D and PfGF. |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7422 |
| Gene Symbol | VEGFA |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse VEGF-A, Biotin-Conjugate anti-mouse VEGF-A polyclonal antibody, Streptavidin-HRP, Mouse VEGF-A Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Diluent, Conjugate Diluent, Concentrate 20x, Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
