Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse VEGFR3/Flt-4 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB009570 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009570 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009570 Supplier Novus Biologicals™ Supplier No. NBP278876

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This kit recognizes Mouse VEGFR3/Flt4 in samples.

No significant cross-reactivity or interference between Mouse VEGFR3/Flt4 and analogues was observed.

Specifications

Accession Number P35917, P35917
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 2324
Gene Symbol FLT4
Host Species Mouse
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target VEGFR3/Flt-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.