Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Mouse Ventilator
SDP

Stoelting™ Mouse Ventilator features a unique design that ensures accuracy, durability and quietness of operation

$10750.00 - $11840.00

Specifications

For Use With (Application) Neuroscience
View More Specs

Products 2
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
10-000-626
SDP
View Documents
Stoelting™
50095
Ventilator 0.5 mL
Each for $10,750.00
Only null left
Add to Cart
 
10-001-068
SDP
View Documents
Stoelting™
500941
Ventilator, Synchronized start/stop 1 mL
Each for $11,840.00
Only null left
Add to Cart
 
Description

Description

  • Unique design, compact and reliable
  • Quiet operation
  • Negligible R.F. broadcasting
  • Ideal for use with mice and also suitable for small birds and perinatal rats
  • Valve mechanism features nearly zero dead space and superb durability with no leakage. Expired air may be channeled for analysis when used with anesthetic gases.
  • Stroke rate is displayed on an LED readout

Principle of Operation
A unique variable stroke linkage mechanism, very similar in principle to the one used in Basile's larger respirators, operates the piston. The reciprocating motion is adjusted and transmitted to the piston by rods and articulated joints only, which leads to minimal wear, no backlash, silent operation and exact stroke reproducibility.

Easily Adjustable Stroke Volume
The stroke volume is adjustable whether the ventilator is on or off. For the most convenient and accurate stroke volume control available, the knob and scale remain stationary, even when the piston is in motion. The valve mechanism on these ventilators features nearly zero dead space and superb durability with no leakage. Expired air may be channeled for analysis and/or recycled, when used with anesthetic gases.

Digital Stroke Rate Display
The stroke rate (breaths per minute) is feedback controlled from 50 to 300 strokes/min and displayed on an LED digital readout. A connection square of four ports (pictured at right) include intake, out to animal, return from animal and exhaust.

Optional controlled pause feature available

Specifications

Specifications

Neuroscience
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.