Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse VSTM2A (NP_001277468) VersaClone cDNA
SDP

Catalog No. RDC2283 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2283 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2283 Supplier R&D Systems™ Supplier No. RDC2283
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Mouse VSTM2A (NP_001277468) VersaClone cDNA plays a role in the regulation of the early stage of white and brown preadipocyte cell differentiation.

The Mouse VSTM2A (NP_001277468) VersaClone cDNA clone contains the complete ORF for VSTM2A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VSTM2A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene VSTM2A
Accession Number NP_001277468
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 772bp
pH 8.5
Gene ID (Entrez) 222008
Gene Alias MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2 A, V-set and transmembrane domain-containing protein 2 A, VSTM2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.