Learn More
Description
- Also known as: E130203B14Rik
- VSTM4 is a type I transmembrane protein in the Immunoglobulin Superfamily.
- A portion of the ECD is cleaved into a 50 aa peptide. This peptide (Peptide Lv) is thought to enhance L-type voltage-gated calcium channel (L-VGCC) currents in retinal photoreceptors.
- The VSTM4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | VSTM4 |
| Accession Number | NP_848906 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 973 bp |
| Gene ID (Entrez) | 196740 |
| Gene Alias | C10orf72, FLJ31737, hypothetical protein LOC196740, V-set and transmembrane domain containing 4 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
