Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Wnt-3a and Inhibitor IWR Set Protein

Catalog No. PSM1324
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSM1324 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PSM1324 Supplier R&D Systems™ Supplier No. PSM1324
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Mouse Wnt-3a and Inhibitor IWR Set Protein is derived from CHO. The Mouse Wnt-3a and Inhibitor IWR Set Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 89780
Quantity 1 Set
Source Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a
Common Name Wnt-3a
Gene Symbol Wnt3a
Biological Activity Recombinant Mouse Wnt-3a induces beta-catenin responsive Topflash activation in HEK293T cells. The specific Wnt pathway inhibitor, endo-IWR 1, inhibits Topflash activity from a 500ng/mL dose of Recombinant Mouse Wnt-3a with a typical ED50 < 0.2μM. The less active diasteriomer exo-IWR 1 Wnt inhibitor shows minor inhibitory activity in this assay.Recombinant Mouse Wnt-3a induces alkaline phosphatase production in the MC3T3-E1 mouse pre-osteoblast cell line. The ED50 for this effect is typically ≤ 5ng/mL. The specific Wnt pathway inhibitor, endo-IWR 1, inhibits alkaline phosphatase production induced from a 10ng/mL dose of Recombinant Mouse Wnt-3a with a typical ED50 < 0.8μM. The less active diasteriomer exo-IWR 1 Wnt inhibitor shows a > 5μM ED50 with a Recombinant Mouse Wnt-3a dose of 10ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.