Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Wnt-9b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB36691100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB36691100 100 μg
MAB36691SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB36691100 Supplier R&D Systems Supplier No. MAB36691100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Wnt-9b Monoclonal specifically detects Wnt-9b in Mouse samples. It is validated for Neutralization.

Specifications

Antigen Wnt-9b
Applications Neutralization
Classification Monoclonal
Clone 964742
Conjugate Unconjugated
Dilution Neutralization 125-750 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O35468
Gene Alias Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, Wnt-14b, WNT14Bprotein Wnt-9b, Wnt-15, WNT15Protein Wnt-15, Wnt9b
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Wnt-9b Met1-Arg359 Accession # O35468
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7484
Test Specificity Detects mouse Wnt-9b in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.