Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit

Catalog No. FCSTM20
Change view
Click to view available options
Quantity:
1 Kit
19 Plex
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCSTM20 1 Kit
FCSTM2019 19 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FCSTM20 Supplier R&D Systems™ Supplier No. FCSTM20

Please contact your sales representative for ordering assistance.

This item is not returnable. View return policy

Specifications

Quantity 1 Kit
Species Mouse
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.