Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit
SDP

Catalog No. FCSTM2019 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
11 Plex
12 Plex
19 Plex
45 Plex
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCSTM2019 19 Plex
FCSTM2045 45 Plex
FCSTM2011 11 Plex
FCSTM2012 12 Plex
FCSTM20 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. FCSTM2019 Supplier R&D Systems™ Supplier No. FCSTM2019
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Quantity 19 Plex
Species Mouse
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.