Learn More
Biomatik Corporation Mouse X-Ray Repair Cross Complementing 6 (XRCC6) ELISA Kit
Supplier:  Biomatik Corporation EKU08246
Encompass_Preferred
Mouse X-Ray Repair Cross Complementing 6 (XRCC6) ELISA Kit for detecting XRCC6 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.107ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23475. Gene ID: 14375. Target Synonyms: Ku70; CTC75; CTCBF; G22P1; ML8; TLAA; Lupus Ku Autoantigen Protein P70; ATP-Dependent DNA Helicase 2 Subunit 1; CTC box-binding factor 75 kDa; Thyroid-lupus autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.