Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse YM1/Chitinase 3-like 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2446SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2446SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2446SP Supplier R&D Systems Supplier No. AF2446SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

YM1/Chitinase 3-like 3 Polyclonal specifically detects YM1/Chitinase 3-like 3 in Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen YM1/Chitinase 3-like 3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35744
Gene Alias AI505981, Chi3l3, Chitinase 3 like 3, Chitinase 3-like 3, ECF-L, Ym1
Gene Symbols CHI3L2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse YM1/Chitinase 3-like 3 Tyr22-Tyr398 Accession # O35744
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1117
Test Specificity Detects mouse YM1/Chitinase 3-like 3 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.