Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moxalactam sodium salt, 50 mg/mL in distilled water, sterile-filtered, Thermo Scientific™

Catalog No. AAJ67056XF
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67056XF 1 mL
AAJ670568EQ 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67056XF Supplier Thermo Scientific Chemicals Supplier No. J67056XF
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64953-12-4
Molecular Formula C20H18N6Na2O9S
Molecular Weight (g/mol) 564.437
MDL Number MFCD03427565
InChI Key GRIXGZQULWMCLU-GDUWRUPCSA-L
PubChem CID 12856838
IUPAC Name disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+]

Specifications

Type Moxalactam Sodium Salt
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Chemical Name or Material Moxalactam sodium salt
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 265-288-4
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.