Learn More
Description
- Zirconium Oxide-coated ceramic grinding spheres
- Chemically inert
- Component of Lysing Matrix A and M tubes
Specifications
Specifications
| Bead Type | Zirconium oxide coated ceramic |
| Content And Storage | Room Temperature |
| Sample Type | Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores |
| For Use With (Application) | Break hard/brittle samples |
| Product Type | Bulk Matrix |
| Quantity | 500 Pk. |
| Sample Volume | Unlimited |
| Synonym | 1/4 in. Zirconium Oxide Banded Satellite for Grinding |
Safety and Handling
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.