Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 1/4 in. Ceramic Spheres

Catalog No. 116540424 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Pk.
1000 Pk.
50 Pk.
500 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-654-0424 100 Pk.
MP116540034 50 Pk.
11-654-0422 1000 Pk.
11-654-0412 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-654-0424 Supplier MP Biomedicals™ Supplier No. 116540424
Add to Cart
Add to Cart

Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Zirconium oxide coated ceramic
Content And Storage Room Temperature
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
For Use With (Application) Break hard/brittle samples
Product Type Bulk Matrix
Quantity 100 Pk.
Sample Volume Unlimited
Synonym 1/4 in. Zirconium Oxide Banded Satellite for Grinding

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.