Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2XYT-Broth Agar, Capsules

Catalog No. MP113012231 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113012231 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113012231 Supplier MP Biomedicals™ Supplier No. 113012231
Add to Cart
Add to Cart

Optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages

  • Use 46 capsules per liter
  • Contents/Liter: 2XYT, 15g agar

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™ FDA
Content And Storage 15°C to 30°C
Format Capsule
Product Type Microbiology Media
Formulation Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L, Agar-B 15g/L
For Use With (Application) Growth and maintenance of M13 phage and other filamentous ssDNA bacteriophages
Quantity 5000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.