Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Acid Phosphatase, Wheat Germ

Catalog No. ICN15387694 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15387694 5 g
ICN15387690 500 mg
ICN15387691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15387694 Supplier MP Biomedicals™ Supplier No. 0215387694
Add to Cart
Add to Cart

  • Activity 20units/mg

TRUSTED_SUSTAINABILITY

Specifications

Concentration Activity: 0.5 U/mg solid; 0.55 U/mg protein
Color Brown
Purity 90%
pH 4 to 7
For Use With (Application) Catalyzes the hydrolysis of Monoesters and Anhydrides of Phosphoric Acid
Content And Storage -20°C
Source Wheat germ
Quantity 5 g
Product Type Acid Phosphatase
Molecular Weight (g/mol) 58
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.