Learn More
MP Biomedicals™ AIN-93G Diet, Pellet
For growth, pregnancy and lactational phases of rodents
Supplier:  MP Biomedicals™ 0296039910
 View more versions of this product
Catalog No. ICN96039910
Only null left
Add to Cart
Description
Description
For sixteen years, the American Institute of Nutrition Rodent Diets, AIN-76 and AIN-76A, have been used extensively around the world. Because of numerous nutritional and technical problems encountered with the diet during this period, it was revised.
Two new formulations were derived:
  • AIN-93 g for growth, pregnancy and lactation
  • AIN-93M for adult maintenance
Some major differences in the new formulation of AIN-93 g compared with AIN-76A are as follows:
  • 7 g Soybean oil/100 g diet was substituted for 5 g corn oil/100 g diet to increase the amount of linolenic acid
  • Cornstarch was substituted for sucrose
  • The amount of phosphorus was reduced to help eliminate the problem of kidney calcification in female rats
  • L-cystine was substituted for DL-methionine as the amino acid supplement for casein, known to be deficient in the sulfur amino acids
  • Manganese concentration was lowered to one fifth the amount in the old diet
  • The amounts of Vitamin E, Vitamin K and Vitamin B12 were increased
  • Molybdenum, silicon, fluoride, nickel, boron, lithium and vanadium were added to the mineral mix
For the AIN-93M maintenance diet:
  • The amount of fat was lowered to 40 g/kg diet from 70 g/kg diet and the amount of casein to 140 g/kg from 200 g/kg in the AIN-93 g diet
Because of a better balance of essential nutrients, the AIN-93 diets may prove to be a better choice than AIN-93A for long-term as well as short-term studies with laboratory rodents. J. Nutr., v. 123: 1939-1951 (1993).
  • AIN-93 g diet is available in pelleted form
  • AIN-93 g diet is used for growth, pregnancy and lactational phases of rodents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Pellet
10 kg
4°C
Casein 20%, Dextrinized Cornstarch 13.2%, Sucrose10%, Corn Starch 39.7486%, Alphacel, Non-Nutritive Bulk 5%, Soybean Oil 7%, AIN-93M Mineral Mix 3.5%, L-Cystine 0.3%, AIN-93-VX Vitamin Mix 1%, Choline bitartrate 0.25%, tert-Butylhydroquinone 0.0014%
Growth, pregnancy and lactational phases of Rodents
Rodent
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.