Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin 35% Solution

Catalog No. ICN810063 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN810063 1 L
ICN810061 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN810063 Supplier MP Biomedicals™ Supplier No. 08810063
Add to Cart
Add to Cart

Globulin free albumins are suitable for use in applications where no other proteins should be present

Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.

  • CAS Number: 9048-46-8
  • MDL Number: MFCD00130384
  • Format: Solution
  • Sterile albumins
  • Concentration: About 35g/dl

For immunoassays and mammalian cell culture. Used to solubilize lipids and is also used as a blocking agent in Western blots or ELISA applications.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 34.4g/dL
For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) ∽66
Name Bovine Albumin 35% Solution
pH Range 7.2
Quantity 1 L
Storage Requirements 0° to 5°C
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.