Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine 30%, Cap Free Sterile

Catalog No. ICN810133 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN810133 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN810133 Supplier MP Biomedicals™ Supplier No. 08810133
Add to Cart
Add to Cart

Sterile

Albumin, Bovine, [9048-46-8]

  • F.W. 66338
  • 30% Solution
  • Cap Free

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) 66338
Name Bovine Albumin
pH Range 7.4
Quantity 50 mL
Source Bovine Blood
Storage Requirements 2° to 8°C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.