Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, RIA Grade

Catalog No. ICN10503325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10503325 25 g
ICN10503310 10 g
ICN10503380 100 g
ICN10503390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10503325 Supplier MP Biomedicals™ Supplier No. 0210503325
Add to Cart
Add to Cart

Prepared by heat shock fractionation

  • Low fatty acids, IgG and hormones
  • Purity: >98%
  • pH: 7
  • Protease-free

Mfr. No. 180537 may substitute

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Western Blot
Molecular Weight (g/mol) 66.338
Name Bovine Serum Albumin
pH Range 6.8 to 7.2
Quantity 25 g
Source Bovine Serum
Storage Requirements 4°C
Form Crystalline
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.