Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Alcohol Dehydrogenase, From Yeast

Catalog No. ICN15143025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15143025 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15143025 Supplier MP Biomedicals™ Supplier No. 0215143025

Specifications

Concentration ≥300 U/mg
Color Yellow
pH 8.6 to 9
For Use With (Application) Enzymatic determination of low concentrations of Ethanol in aqueous samples
Content And Storage 0°C
Source Yeast
Quantity 25 mg
Product Type Alcohol Dehydrogenase
Molecular Weight (g/mol) 141000
Inhibitors Heavy Metals, -SH reagents
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.